missing translation for 'onlineSavingsMsg'
Learn More
Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals Product Code.: 10397841

Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals

Product Code. 10397841
250 mL, Glass Bottle
Click to view available options
Quantity:
250 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10397841

Brand: Thermo Scientific Acros 327182500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 18.4°C
CAS 67-68-5
Density 1.1000g/mL
Boiling Point 189.0°C
Quantity 250 mL
Flash Point 87°C
Packaging Glass Bottle
Physical Form Liquid at 20°C
Refractive Index 1.4770 to 1.4800
Assay Percent Range 99.8+%
Residue after Evaporation 0.001% max.
Specific Gravity 1.1
Molecular Formula C2H6OS
Linear Formula (CH3)2SO
MDL Number MFCD00002089
Viscosity 1.987 mPa.s (25°C)
Beilstein 01, IV, 1277
Water 0.1% max. (Coulometric)
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methylsulfinylmethane
Molecular Weight (g/mol) 78.13
PubChem CID 679
ChEBI CHEBI:28262
Formula Weight 78.13
Percent Purity ≥99.8%
Grade Molecular Biology
Chemical Name or Material Methyl sulfoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464015 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number327182500
Lot NumberA0445514 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number327182500
Lot NumberA0458087 Certificate TypeCertificate of Analysis Date30/04/2024 Catalog Number327182500
Lot NumberA0451924 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number327182500
Lot NumberA0397417 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number327182500
Product Identifier
  • Methyl sulfoxide
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    5

    Minutes

    11

    Seconds

    11

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.