missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Product Code. 10438920
250 mL, Glass Bottle
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Absorbance 0.01 max. at 360nm, 0.05 max. at 320nm, 0.1 max. at 300nm, 0.2 max. at 290nm, 0.3 max. at 280nm, 0.5 max. at 270nm
Decomposition Information >190°C
Density 1.1000g/mL
Boiling Point 189.0°C
Quantity 250 mL
Flash Point 87°C
Packaging Glass Bottle
Physical Form Liquid at 20°C
Refractive Index 1.4770 to 1.4800
Assay Percent Range 99.9%
Residue after Evaporation 0.005% max.
Specific Gravity 1.1
Linear Formula (CH3)2SO
Vapor Pressure Vapor Pressure: 0.55mbar at 20°C
Viscosity 1.987 mPa.s (25°C)
Beilstein 01, IV, 1277
Water 0.05% max.
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity 99.9%
Grade Spectroscopy
Chemical Name or Material Methyl sulfoxide, 99.9%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468347 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number167850025,167852500,167851000,167850010
Lot NumberA0456074 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number167850025,167852500,167851000,167850010
Lot NumberA0466065 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number167850025,167852500,167850010,167851000
Lot NumberA0465070 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog Number167850025,167852500,167850010,167851000
Lot NumberA0450705 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number167850025,167852500,167850010,167851000
Product Identifier
  • Methyl sulfoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.