missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-Tert-Butyl Ether, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
HDPE Plastic Bottle
Metal can
Plastic Coated Glass bottle
Description
C5H12O, CAS Number-1634-04-4, methyl t-butyl ether, 2-methyl-2-methoxypropane, methyl tert-butyl ether, propane, 2-methoxy-2-methyl, methyl-t-butyl ether, mtbe, methyl tertiary-butyl ether, methyl-tert-butyl ether, tert-butyl methyl ether, t-butyl methyl ether, 5L, 54 deg.C, CHEBI:27642, Colorless

Specifications
Specifications
Melting Point | -110°C |
Color | Colorless |
Boiling Point | 54°C |
Packaging | HDPE Plastic Bottle |
Quantity | 5 L |
Formula Weight | 88.15g/mol |
Vapor Pressure | 268mbar at 20°C |
Physical Form | Liquid |
Safety and Handling


Product Identifier
- tert-Butyl methyl ether
- Danger
- Skin corrosion/irritation Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl-Tert-Butyl Ether, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction