missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiazole-2-carboxylate, 95%

Product Code. 15490219
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15490219

Brand: Thermo Scientific Alfa Aesar H63496.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55842-56-3 | C5H5NO2S | 143.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55842-56-3
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD18075580
InChI Key YRMZUOPMHMPNPG-UHFFFAOYSA-N
PubChem CID 7567660
IUPAC Name methyl 1,3-thiazole-2-carboxylate
SMILES COC(=O)C1=NC=CS1

Specifications

Quantity 5 g
Synonym methyl thiazole-2-carboxylate, methyl 2-thiazolecarboxylate, 2-thiazolecarboxylic acid methyl ester, methyl2-thiazolecarboxylate
Formula Weight 143.16
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl thiazole-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.