Learn More
Methyl (trimethylsilyl)propiolate, 98%, Thermo Scientific Chemicals
£71.46 - £1112.54
Chemical Identifiers
CAS | 42201-71-8 |
---|---|
Molecular Formula | C7H12O2Si |
Molecular Weight (g/mol) | 156.256 |
MDL Number | MFCD04039783 |
InChI Key | XXGHAMBKHIDZDC-UHFFFAOYSA-N |
Synonym | methyl trimethylsilyl propiolate, methyl 3-trimethylsilylpropynoate, methyl 3-trimethylsilyl propiolate, methyl 3-trimethylsilyl-2-propynoate, acmc-20akhx, methyl 3-trimethylsilylpropiolate, methyl 3-trimethylsilyl propynoate, trimethylsilylpropiolic acid methyl ester, trimethylsilyl propiolic acid methyl ester, methyl 3-trimethylsilyl-2-propynoate #Show More |
PubChem CID | 560731 |
IUPAC Name | methyl 3-trimethylsilylprop-2-ynoate |
SMILES | COC(=O)C#C[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470778
|
Thermo Scientific Alfa Aesar
B25245.03 |
1 g |
£71.46
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480778
|
Thermo Scientific Alfa Aesar
B25245.06 |
5 g |
£277.38
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490778
|
Thermo Scientific Alfa Aesar
B25245.14 |
25 g | N/A | ||||||
Description
Methyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Chemical Identifiers
42201-71-8 | |
156.256 | |
XXGHAMBKHIDZDC-UHFFFAOYSA-N | |
560731 | |
COC(=O)C#C[Si](C)(C)C |
C7H12O2Si | |
MFCD04039783 | |
methyl trimethylsilyl propiolate, methyl 3-trimethylsilylpropynoate, methyl 3-trimethylsilyl propiolate, methyl 3-trimethylsilyl-2-propynoate, acmc-20akhx, methyl 3-trimethylsilylpropiolate, methyl 3-trimethylsilyl propynoate, trimethylsilylpropiolic acid methyl ester, trimethylsilyl propiolic acid methyl ester, methyl 3-trimethylsilyl-2-propynoate #Show More | |
methyl 3-trimethylsilylprop-2-ynoate |
Specifications
71°C to 75°C (11mmHg) | |
C7H12O2Si | |
MFCD04039783 | |
methyl trimethylsilyl propiolate, methyl 3-trimethylsilylpropynoate, methyl 3-trimethylsilyl propiolate, methyl 3-trimethylsilyl-2-propynoate, acmc-20akhx, methyl 3-trimethylsilylpropiolate, methyl 3-trimethylsilyl propynoate, trimethylsilylpropiolic acid methyl ester, trimethylsilyl propiolic acid methyl ester, methyl 3-trimethylsilyl-2-propynoate # | |
COC(=O)C#C[Si](C)(C)C | |
156.256 | |
156.25 | |
1.44 |
42201-71-8 | |
(CH3)3SiC≡CCO2CH3 | |
UN3272 | |
XXGHAMBKHIDZDC-UHFFFAOYSA-N | |
methyl 3-trimethylsilylprop-2-ynoate | |
560731 | |
98% | |
Methyl (trimethylsilyl)propiolate |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.