missing translation for 'onlineSavingsMsg'
Learn More
Methyl Vinyl Ketone, Stabilized ca 95%, Thermo Scientific Chemicals Product Code.: 16221612
compliance-icons compliance-icons compliance-icons compliance-icons

Methyl Vinyl Ketone, Stabilized ca 95%, Thermo Scientific Chemicals

Product Code. 16221612
100 mL, Glass bottle
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 16221612

Brand: Thermo Scientific Acros 128001000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-94-4, 75-05-8, 64-19-7, 123-31-9, 7732-18-5
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00008777
InChI Key FUSUHKVFWTUUBE-UHFFFAOYSA-N
Synonym methyl ethenyl ketone, butenone, 2-butenone, methylene acetone, methyl vinyl ketone, vinyl methyl ketone, 3-butene-2-one, acetyl ethylene, methylvinylketon, 3-buten-2-one
PubChem CID 6570
ChEBI CHEBI:48058
IUPAC Name but-3-en-2-one
SMILES CC(=O)C=C

Specifications

Color Brown to Colorless
Density 0.8640 g/mL
Boiling Point 80.0°C
Flash Point -7°C
Infrared Spectrum Authentic
Assay Percent Range ca. 95%
Packaging Glass bottle
Linear Formula H2C=CHCOCH3
Refractive Index 1.4100 to 1.4120
Quantity 100 mL
Fieser 01,697; 02,283; 05,464; 06,407; 07,247; 10,272; 12,329; 13,193
Specific Gravity 0.864
Stabilizer 0.7% acetonitrile max., 0.4 to 1.0% acetic acid, approx. 0.3% hydroquinone
Formula Weight 70.09
Percent Purity ca. 95%
Physical Form Liquid
Chemical Name or Material Methyl vinyl ketone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0393754 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number128000010,128001000,12800017,128000050,12800082,128000025
Lot NumberA0396192 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number128000010,128001000,12800017,128000050,12800082,128000025
Lot NumberA0408713 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number128000010,128001000,12800017,128000050,12800082,128000025
Lot NumberA0410305 Certificate TypeCertificate of Analysis Date02/10/2021 Catalog Number128000010,128001000,12800017,128000050,12800082,128000025
Lot NumberA0414262 Certificate TypeCertificate of Analysis Date30/07/2021 Catalog Number128000010,128001000,12800017,128000050,12800082,128000025
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl vinyl ketone, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl Vinyl Ketone, Stabilized ca 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.