missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Violet 2B
Click to view available options
Quantity:
50 g
250 g
Description
Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton?s crystal violet-iodine technique for chromatin and nucleoli; Lieb?s amyloid stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton′s crystal violet-iodine technique for chromatin and nucleoli; Lieb′s amyloid stain.
Solubility
Soluble in water.
Notes
Transition interval: pH 0.0 (yellow) to pH 1.6 (blue)
Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton′s crystal violet-iodine technique for chromatin and nucleoli; Lieb′s amyloid stain.
Solubility
Soluble in water.
Notes
Transition interval: pH 0.0 (yellow) to pH 1.6 (blue)
Specifications
Solubility Information | Soluble in water. |
Formula Weight | 393.96 |
Odor | Odorless |
Quantity | 50 g |
Chemical Name or Material | Methyl Violet 2B |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'