Learn More
Thermo Scientific Chemicals Methyl Violet 2B, pure, indicator grade
£26.73 - £87.44
Chemical Identifiers
CAS | 8004-87-3 |
---|---|
Molecular Formula | C24H28ClN3 |
MDL Number | MFCD00012568 |
Synonym | Basic Violet 1, C.I. 42535 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10073120
|
Thermo Scientific Chemicals
228371000 |
100 g | Glass bottle |
£26.73
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10125050
|
Thermo Scientific Chemicals
228375000 |
500 g | Glass bottle |
£87.44
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
8004-87-3 | |
C24H28ClN3 | |
MFCD00012568 | |
Basic Violet 1, C.I. 42535 |
Specifications
137.0°C | |
1 to 1.25 | |
MFCD00012568 | |
Basic Violet 1, C.I. 42535 | |
Authentic | |
393.95 | |
Pure | |
Green | |
Methyl violet 2B |
8004-87-3 | |
C24H28ClN3 | |
13, 755 | |
Solubility in water: soluble. Other solubilities: soluble in conc. sulfuric acid and nitric acid, soluble in chloroform, partially soluble in alcohol and glycerol | |
580 to 586nm (in 50:50 IMS:water) | |
10% max. (105°C) | |
75% min. (on dry substance) | |
Crystalline Powder With Metal Luster |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'rtecsNumber' : GC8425000
missing translation for 'tsca' : TSCA
- Methyl Violet 2B, high purity biological stain
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.