missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet, Pure, Solution 1%, C.I.2555, Indicator Grade, Fisher Chemical

Product Code. 10615502
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

MDL Number 11750
Packaging Amber glass bottle
Color Green
Quantity 500 mL
Physical Form Liquid
compliance-icons
Product Identifier
  • Methyl violet solution 1% indicator
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : C.I. Basic Violet 3,Michler's ketone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'