missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Yellow, pure, indicator grade

Product Code. 10170030
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-11-7
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.30
MDL Number MFCD00008308
InChI Key JCYPECIVGRXBMO-UHFFFAOYSA-N
Synonym 4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2
PubChem CID 6053
ChEBI CHEBI:17903
IUPAC Name N,N-dimethyl-4-phenyldiazenylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 111.0°C to 117.0°C
Linear Formula (CH3)2NC6H4N=NC6H5
Solubility Information Solubility in water: 13.6 mg/l. Other solubilities: soluble ca. 140g/L in acetone, soluble in benzene, chloroform, diethyl ether,, ethanol, petroleum ether, vegetable oils, alcohol,, ether, strong mineral acids and soils
Infrared Spectrum Authentic
Formula Weight 225.29
Loss on Drying 3% max.
Grade Pure
Packaging Glass bottle
Quantity 500 g
Physical Form Powder
Chemical Name or Material Methyl Yellow
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl Yellow, indicator grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N,N-dimethyl-4-(phenylazo)benzenamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'