missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Methyl Yellow, pure, indicator grade Product Code.: 10219183

Thermo Scientific Chemicals Methyl Yellow, pure, indicator grade

Product Code. 10219183
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10219183

Brand: Thermo Scientific Chemicals 151400250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-11-7
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.30
MDL Number MFCD00008308
InChI Key JCYPECIVGRXBMO-UHFFFAOYSA-N
Synonym 4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2
PubChem CID 6053
ChEBI CHEBI:17903
IUPAC Name N,N-dimethyl-4-phenyldiazenylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 111.0°C to 117.0°C
Linear Formula (CH3)2NC6H4N=NC6H5
Solubility Information Solubility in water: 13.6 mg/l. Other solubilities: soluble ca. 140g/L in acetone, soluble in benzene, chloroform, diethyl ether,, ethanol, petroleum ether, vegetable oils, alcohol,, ether, strong mineral acids and soils
Infrared Spectrum Authentic
Formula Weight 225.29
Loss on Drying 3% max.
Grade Pure
Packaging Glass bottle
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl Yellow
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466731 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number151405000,151401000,15140148,151400250,15140122
Lot NumberA0452328 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number151405000,151401000,15140148,151400250,15140122
Lot NumberA0420370 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number151405000,151401000,15140148,151400250,15140122
Lot NumberA0409951 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number151405000,151401000,15140148,151400250,15140122
Lot NumberA0408604 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number151405000,151401000,15140148,151400250,15140122
compliance-icons compliance-icons
Product Identifier
  • Methyl Yellow, indicator grade
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash hands and any exposed skin thoroughly after handling.
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Methyl Yellow, pure, indicator grade >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.