missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Methylate Reagent (DMFDMA)

For easy, effective preparation of methyl esters from fatty acids and amino acids use Thermo Scientific™ Methylate Reagent (DMFDMA).

Brand:  Thermo Scientific™ TS-49350


Product Code. 11527690

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

For easy, effective preparation of methyl esters from fatty acids and amino acids use Thermo Scientific™ Methylate Reagent (DMFDMA). Reactions are typically complete upon dissolution (except for long-chain solid acids) and quantitative yields are obtained when reagent and sample are injected without prior mixing. No water washing, extraction, or concentration of derivatives is required and no water is formed. Methylate Reagent (DMFDMA) contains 2 mEq/mL in pyridine and is supplied in ready-to-use Thermo Scientific™ Hypo-Vial™ Sample Storage Vials.

Advantages for preparation of methyl esters for gas chromatography:

  • Speed: the reaction is complete upon dissolution (except long chain solid acids)
  • No water washing, extraction or concentration of derivatives required
  • No water formed
  • Quantitation: quantitative yields are obtained when the reagent and sample are injected without prior mixing
  • Convenient: ready-to-use reagent contains 2mEq/mL pyridine

Ordering Alerts:

Packaged in Hypo-Vial Sample Storage Vial.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC, Columns 0.53mm I.D., 0.8 mm Ferrule I.D.
Room Temperature
65 - 80, 20 - 25 %
Flammable, Harmful, Toxic
Pyridine, 1,1-Dimethoxy-N,N-dimethyl-methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • methyl 8 reagent

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P271-Use only outdoors or in a well-ventilated area.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methanamine, 1,1-dimethoxy-N,N-dimethyl-,pyridine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.