missing translation for 'onlineSavingsMsg'
Learn More

Methylated Spirit Mineralised, Technical, Fisher Chemical™

Catalog Number 11412874
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle
This item is not returnable. View return policy
64-17-5
C2H6O
46.07
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
This item is not returnable. View return policy
64-17-5
C2H6O
46.07
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236

Methylated Spirit Mineralised, Technical

TRUSTED_SUSTAINABILITY
Color Purple
Packaging Amber glass bottle
Quantity 2.5 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Methylated spirit mineralised
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Denatonium benzoate, Methyl ethyl ketone, Isopropyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'