missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Methylation Smittel TMSH
£144.00 - £273.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10123013
|
Macherey-Nagel™
701520.110 |
10 mL |
£144.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10389991
|
Macherey-Nagel™
701520.201 |
1 mL |
£273.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- TMSH, 0,2 M, 1x10 mL
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : methanol, TMSH