missing translation for 'onlineSavingsMsg'
Learn More

Methylcobalamin hydrate, 97%, Thermo Scientific Chemicals

Product Code. 11449113
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.

Solubility
Partially soluble in cold water, hot water.

Notes
Store at -20°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288315-09-3
Molecular Formula C63H91CoN13O14P-
Molecular Weight (g/mol) 1344.405
MDL Number MFCD00149221
InChI Key QXOMMGINPOYKPR-YHKGLSPZSA-L
Synonym methylcobalamin
PubChem CID 134128059
IUPAC Name carbanide;cobalt(2+);[(2S,3R,4S,5R)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2S)-1-[3-[(1S,2S,3S,5Z,7R,10Z,12R,13R,15Z,17R,18R,19S)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-o
SMILES [CH3-].CC1=CC2=C(C=C1C)N(C=N2)C3C(C(C(O3)CO)OP(=O)([O-])OC(C)CNC(=O)CCC4(C(C5C6(C(C(C(=C(C7=NC(=CC8=NC(=C(C4=N5)C)C(C8(C)C)CCC(=O)N)C(C7(C)CC(=O)N)CCC(=O)N)C)[N-]6)CCC(=O)N)(C)CC(=O)N)C)CC(=O)N)C)O.[Co+2]

Specifications

Quantity 500 mg
Solubility Information Partially soluble in cold water,hot water.
Formula Weight 1344.41 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Methylcobalamin hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'