Learn More
Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals
£35.00 - £190.00
Chemical Identifiers
CAS | 108-87-2 |
---|---|
Molecular Formula | C7H14 |
Molecular Weight (g/mol) | 98.19 |
MDL Number | MFCD00001497 |
InChI Key | UAEPNZWRGJTJPN-UHFFFAOYSA-N |
Synonym | cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan |
PubChem CID | 7962 |
IUPAC Name | methylcyclohexane |
SMILES | CC1CCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10091200
|
Thermo Scientific Acros
126580010 |
1 L | Glass Bottle |
£35.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031180
|
Thermo Scientific Acros
126580025 |
2.5 L | Glass Bottle |
£66.90
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10225621
|
Thermo Scientific Acros
126580050 |
5 L | Plastic drum |
£112.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10367701
|
Thermo Scientific Acros
126580100 |
10 L | Plastic drum |
£190.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-87-2 | |
98.19 | |
UAEPNZWRGJTJPN-UHFFFAOYSA-N | |
7962 | |
CC1CCCCC1 |
C7H14 | |
MFCD00001497 | |
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan | |
methylcyclohexane |
Specifications
108-87-2 | |
0.7700g/mL | |
−3°C | |
98.5% min. (GC) | |
C6H11CH3 | |
MFCD00001497 | |
15, 6118 | |
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan | |
UAEPNZWRGJTJPN-UHFFFAOYSA-N | |
methylcyclohexane | |
7962 | |
99% | |
Liquid |
-126.3°C | |
101.0°C | |
Authentic | |
C7H14 | |
1.4210 to 1.4230 | |
05, 29 | |
0.77 | |
Solubility in water: 0.1g/L (20°C). Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,ether,ethanol | |
CC1CCCCC1 | |
98.19 | |
98.19 | |
Extra Pure | |
Methylcyclohexane, 99% |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-624-3
missing translation for 'rtecsNumber' : GV6125000
missing translation for 'tsca' : TSCA
- Methylcyclohexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
RUO – Research Use Only