missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcyclohexane, 99%, Spectrophotometric Grade, Honeywell Riedel-de Haën™
Spectrophotometric Grade, 99%
£181.86 - £314.08
Chemical Identifiers
CAS | 108-87-2 |
---|---|
Molecular Formula | C7H14 |
Molecular Weight (g/mol) | 98.19 |
MDL Number | MFCD00001497 |
InChI Key | UAEPNZWRGJTJPN-UHFFFAOYSA-N |
Synonym | cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan |
PubChem CID | 7962 |
IUPAC Name | methylcyclohexane |
SMILES | CC1CCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15612800
|
Honeywell Riedel-de Haën
259691-1L |
1 L | Glass bottle |
£181.86
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15622800
|
Honeywell Riedel-de Haën
259691-2L |
2 L | Glass bottle |
£314.08
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
Chemical Identifiers
108-87-2 | |
98.19 | |
UAEPNZWRGJTJPN-UHFFFAOYSA-N | |
7962 | |
CC1CCCCC1 |
C7H14 | |
MFCD00001497 | |
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan | |
methylcyclohexane |
Specifications
108-87-2 | |
0.77g/mL at 25°C | |
−4°C (24.8°F) | |
C6H11CH3 | |
MFCD00001497 | |
505972 | |
UAEPNZWRGJTJPN-UHFFFAOYSA-N | |
methylcyclohexane | |
7962 | |
99% | |
Methylcyclohexane |
-126°C | |
100°C to 102°C | |
C7H14 | |
1.422 | |
UN2296 | |
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan | |
CC1CCCCC1 | |
98.19 | |
98.19g/mol | |
Sprectrophotometric |
Safety and Handling
P210-P301 + P310-P331-P370 + P378-P403 + P235
H225-H304-H315-H336-H410
missing translation for 'dotInformation' : 3/PGII
missing translation for 'einecsNumber' : 203-624-3
missing translation for 'rtecsNumber' : GV6125000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title