Learn More
Methylcyclopentane, 95%, Thermo Scientific Chemicals
£148.00 - £3113.00
Chemical Identifiers
CAS | 96-37-7 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.16 |
MDL Number | MFCD00001382 |
InChI Key | GDOPTJXRTPNYNR-UHFFFAOYSA-N |
Synonym | cyclopentane, methyl, methyl cyclopentane, methylpentamethylene, unii-5g26cc1ask, methyl-cyclopentane, ccris 6058, hsdb 876, 5g26cc1ask, un2298 flammable liquid, 3-methylcyclopentyl |
PubChem CID | 7296 |
IUPAC Name | methylcyclopentane |
SMILES | CC1CCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10397361
|
Thermo Scientific Acros
345245000 |
500 mL | Glass bottle |
£148.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10277283
|
Thermo Scientific Acros
345240025 |
2.5 L | Glass bottle |
£435.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10736443
|
Thermo Scientific Acros
345240250 |
25 L | Metal Drum |
£3113.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96-37-7 | |
84.16 | |
GDOPTJXRTPNYNR-UHFFFAOYSA-N | |
7296 | |
CC1CCCC1 |
C6H12 | |
MFCD00001382 | |
cyclopentane, methyl, methyl cyclopentane, methylpentamethylene, unii-5g26cc1ask, methyl-cyclopentane, ccris 6058, hsdb 876, 5g26cc1ask, un2298 flammable liquid, 3-methylcyclopentyl | |
methylcyclopentane |
Specifications
96-37-7 | |
White | |
71.0°C | |
Authentic | |
C6H12 | |
1.4077 to 1.4097 | |
05, 27 | |
cyclopentane, methyl, methyl cyclopentane, methylpentamethylene, unii-5g26cc1ask, methyl-cyclopentane, ccris 6058, hsdb 876, 5g26cc1ask, un2298 flammable liquid, 3-methylcyclopentyl | |
GDOPTJXRTPNYNR-UHFFFAOYSA-N | |
methylcyclopentane | |
7296 | |
84.16 | |
Adhering Crystals |
-142.4°C | |
0.7480g/mL | |
−4°C | |
94% min. (GC) | |
C5H9CH3 | |
MFCD00001382 | |
0.748 | |
Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,diethyl ether,ethanol,petroleum ether | |
CC1CCCC1 | |
84.16 | |
0.505 mPa.s (20°C) | |
95% | |
Methylcyclopentane, 95% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-503-2
missing translation for 'rtecsNumber' : GY4640000
missing translation for 'tsca' : TSCA
- Methylcyclopentane
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only