missing translation for 'onlineSavingsMsg'
Learn More

Methyldiethoxysilane, 97%

Product Code. p-3593465 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10607864

Brand: Thermo Scientific Acros 392260050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2031-62-1 | C5H14O2Si | 134.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2031-62-1
Molecular Formula C5H14O2Si
Molecular Weight (g/mol) 134.25
MDL Number MFCD00026749
InChI Key GAURFLBIDLSLQU-UHFFFAOYSA-N
Synonym diethoxymethylsilane, methyldiethoxysilane, diethoxy methyl silane, silane, diethoxymethyl, diethoxy-methylsilane, diethoxy methyl silicon, diethoxymethylsilane. methylhydrogendiethoxysilane., diethoxymethyl-silan, diethoxy methyl silyl
PubChem CID 6327387
IUPAC Name diethoxy(methyl)silicon
SMILES CCO[Si](C)OCC

Specifications

Color Colorless
Density 0.8290g/mL
Boiling Point 94.0°C to 95.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CH2O)2SI(H)CH3
Refractive Index 1.3740 to 1.3760 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.829
Solubility Information Solubility in water: reacts
Formula Weight 134.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyldiethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyldiethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.