missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyldiphenylphosphine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
50 g
Description
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.064 |
Boiling Point | 160°C (15 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 50 g |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 200.21 |
Percent Purity | 99% |
Chemical Name or Material | Methyldiphenylphosphine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'