missing translation for 'onlineSavingsMsg'
Learn More

Methyle1-methyl-1H-imidazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10550933
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 17289-20-2
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD01567300
InChI Key AKDPLDCXQNEMCL-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-imidazole-5-carboxylate, methyl 1-methylimidazole-5-carboxylate, methyl1-methyl-1h-imidazole-5-carboxylate, 3-methyl-3h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-5-carboxylic acid, 1-methyl-, methyl ester, 1-methyl-1h-imidazole-5-carboxylic acid methyl ester, pubchem9004, acmc-209e5h, 1-methylimidazole-5-carboxylic acid methyl ester, 1h-imidazole-5-carboxylicacid, 1-methyl-, methyl ester
PubChem CID 2736896
IUPAC Name methyl 3-methylimidazole-4-carboxylate
SMILES COC(=O)C1=CN=CN1C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Methyle1-methyl-1H-imidazole-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'