missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-aminothiophene-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10394662
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4651-81-4
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00159547
InChI Key DGGJQLCAYQCPDD-UHFFFAOYSA-N
Synonym methyl 2-amino-3-thenoate, methyl 2-amino-3-thiophenecarboxylate, 2-amino-3-methoxycarbonylthiophene, methyl2-aminothiophene-3-carboxylate, 2-amino-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 2-amino-, methyl ester, zlchem 402, pubchem5551, maybridge1_003714, 2-amino-thiophene-3-carboxylicacidmethylester
PubChem CID 78381
IUPAC Name methyl 2-aminothiophene-3-carboxylate
SMILES COC(=O)C1=C(N)SC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 157.19
Percent Purity 97%
Chemical Name or Material Methyle2-aminothiophene-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'