missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-isothiocyanatobenzoate, 97%, Thermo Scientific™

Product Code. 10743574
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 10743574

Brand: Thermo Scientific Maybridge SB01695EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16024-82-1
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041062
InChI Key UNXVHBOJSCWVCD-UHFFFAOYSA-N
Synonym 2-isothiocyanato-benzoic acid methyl ester, 2-methoxycarbonylphenyl isothiocyanate, 2-methoxycarbonyl phenyl isothiocyanate, benzoic acid,2-isothiocyanato-, methyl ester, benzoic acid, 2-isothiocyanato-, methyl ester, acmc-209dkr, methyl2-isothiocyanatobenzoate, methyl 2-isothiocyanato benzoate, methyl 2-isothiocyanatobenzenecarboxylate, 2-isothiocyanatobenzoic acid methyl ester
PubChem CID 85243
IUPAC Name methyl 2-isothiocyanatobenzoate
SMILES COC(=O)C1=CC=CC=C1N=C=S

Specifications

CAS Min % 97.0
Density 1.2500g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula SCNC6H4CO2CH3
Quantity 10 g
Formula Weight 193.23
Percent Purity 97%
Chemical Name or Material Methyl 2-isothiocyanatobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.