Learn More
Thermo Scientific Maybridge Methyl 3-[(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate, 1g, 97%
Brand: Thermo Scientific Maybridge KM06349DA
Methyl 3-[(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate, 1g, 97%
Code : 31
Additional Details : CAS Number : 175203-23-3 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III