missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-bromothiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog Number 10120652
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
26137-08-6
C6H5BrO2S
221.068
MFCD00173839
PEGSJNCGPSIJOX-UHFFFAOYSA-N
3-bromothiophene-2-carboxylic acid methyl ester, 3-bromo-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-bromo-, methyl ester, 3-bromo-2-thiophenecarboxylic acid methyl ester, methyl 3-bromo-2-thiophenecarboxylate, maybridge3_006386, ksc494q2p, 2-methoxycarbonyl-3-bromothiophene, methyl-3-bromothiophene carboxylate, methyl 3-bromanylthiophene-2-carboxylate
2740074
methyl 3-bromothiophene-2-carboxylate
COC(=O)C1=C(C=CS1)Br
This item is not returnable. View return policy
26137-08-6
C6H5BrO2S
221.068
MFCD00173839
PEGSJNCGPSIJOX-UHFFFAOYSA-N
3-bromothiophene-2-carboxylic acid methyl ester, 3-bromo-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-bromo-, methyl ester, 3-bromo-2-thiophenecarboxylic acid methyl ester, methyl 3-bromo-2-thiophenecarboxylate, maybridge3_006386, ksc494q2p, 2-methoxycarbonyl-3-bromothiophene, methyl-3-bromothiophene carboxylate, methyl 3-bromanylthiophene-2-carboxylate
2740074
methyl 3-bromothiophene-2-carboxylate
COC(=O)C1=C(C=CS1)Br
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.07
Percent Purity 97%
Chemical Name or Material Methyle3-bromothiophene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'