missing translation for 'onlineSavingsMsg'
Learn More
Methyle3-chloro-4-methylthiophene-2-carboxylate, 97%, Thermo Scientific™ Product Code.: 10150592

Methyle3-chloro-4-methylthiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10150592
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10150592

Brand: Thermo Scientific Maybridge GK01297DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-11-8
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00052072
InChI Key BUFSIDWTJVUAER-UHFFFAOYSA-N
Synonym methyl 3-chloro-4-methyl-2-thiophenecarboxylate, 3-chloro-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-chloro-4-methyl-, methyl ester, maybridge1_003874, 2-methoxycarbonyl-3-chloro4-methylthiophene, 2-methoxycarbonyl-3-chloro-4-methylthiophene, methyl 3-chloro-4-methylthiophenecarboxylate, methyl3-chloro-4-methylthiophene-2-carboxylate, methyl 3-chloranyl-4-methyl-thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-chloro-4-methyl-, methyl esterShow MoreShow Less
PubChem CID 2777567
IUPAC Name methyl 3-chloro-4-methylthiophene-2-carboxylate
SMILES CC1=CSC(=C1Cl)C(=O)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.65
Percent Purity 97%
Chemical Name or Material Methyl 3-chloro-4-methylthiophene-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyle3-chloro-4-methylthiophene-2-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.