missing translation for 'onlineSavingsMsg'
Learn More
Methyle3-hydroxybutanoate, 97%, Thermo Scientific™ Product Code.: 10778835

Methyle3-hydroxybutanoate, 97%, Thermo Scientific™

Product Code. 10778835
10 g
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 10778835

Brand: Thermo Scientific Maybridge SB01506EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3976-69-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00063289
InChI Key LDLDJEAVRNAEBW-SCSAIBSYSA-N
Synonym methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester
PubChem CID 2724279
IUPAC Name methyl (3R)-3-hydroxybutanoate
SMILES CC(CC(=O)OC)O

Specifications

CAS Min % 97.0
Density 1.0500g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CH3CH(OH)CH2CO2CH3
Quantity 10 g
Specific Rotation Condition - 21.50 (20.00°C neat)
Formula Weight 118.13
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxybutanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyle3-hydroxybutanoate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.