missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-methyl-4-nitrobenzoate, 97%, Thermo Scientific™

Product Code. 10203572
Click to view available options
Quantity:
1 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 24078-21-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00085640
InChI Key IEFONJKJLZFGKQ-UHFFFAOYSA-N
Synonym benzoic acid, 3-methyl-4-nitro-, methyl ester, 3-methyl-4-nitrobenzoic acid methyl ester, m-toluic acid, 4-nitro-, methyl ester, methyl 3-methyl-4-nitrobenzenecarboxylate, benzoic acid, 3-methyl-4-nitro, methyl ester, methyl3-methyl-4-nitrobenzoate, pubchem2350, acmc-1cfdu, methyl 4-nitro-m-toluate, ksc205m3d
PubChem CID 260927
IUPAC Name methyl 3-methyl-4-nitrobenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material Methyl 3-methyl-4-nitrobenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'