missing translation for 'onlineSavingsMsg'
Learn More
Methyle5-methyl-3-isoxazolecarboxylate, 97%, Thermo Scientific™ Product Code.: 10113264

Methyle5-methyl-3-isoxazolecarboxylate, 97%, Thermo Scientific™

Product Code. 10113264
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10113264

Brand: Thermo Scientific Maybridge CC10422DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 19788-35-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H7NO3
MDL Number MFCD00015895
Quantity 1 g
Synonym methyl 5-methylisoxazole-3-carboxylate, methyl 5-methyl-3-isoxazolecarboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, methyl ester, 5-methyl-3-isoxazolecarboxylic acid methyl ester, methyl 5-methyl-3-isoxazole carboxylate, methyl-5-methyl-3-isoxazole carboxylate, 5-methylisoxazole-3-carboxylic acid methyl ester, 5-methyl-isoxazole-3-carboxylic acid methyl ester, acmc-209xf5, ksc489s1r
InChI Key MVHHQOCEOUNTID-UHFFFAOYSA-N
SMILES COC(=O)C1=NOC(C)=C1
IUPAC Name methyl 5-methyl-1,2-oxazole-3-carboxylate
Molecular Weight (g/mol) 141.13
PubChem CID 88245
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material Methyle5-methyl-3-isoxazolecarboxylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyle5-methyl-3-isoxazolecarboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.