missing translation for 'onlineSavingsMsg'
Learn More

Methyle6-(1-pyrrolidinyl)nicotinate, 97%, Thermo Scientific™

Product Code. 10055262
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 210963-94-3
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD05664433
InChI Key OWOKTZZFJFFUKY-UHFFFAOYSA-N
Synonym methyl 6-pyrrolidinonicotinate, methyl 6-1-pyrrolidinyl nicotinate, methyl 6-pyrrolidin-1-yl nicotinate, methyl 6-pyrrolidin-1-yl pyridine-3-carboxylate, acmc-1cher, methyl 6-pyrrolidine-1-yl nicotinate, methyl 6-pyrrolidine-1-yl nicotinoate, methyl 6-pyrrolidin-1-ylnicotinate, methyl 6-pyrrolidinylpyridine-3-carboxylate, methyl 6-pyrrolidin-1-yl nicotinate;ethyl 6-pyrrolidin-1-yl pyridine-3-carboxylate
PubChem CID 2794748
IUPAC Name methyl 6-pyrrolidin-1-ylpyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N2CCCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material Methyle6-(1-pyrrolidinyl)nicotinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.