missing translation for 'onlineSavingsMsg'
Learn More

Methyl chloroacetate, 98%, Thermo Scientific Chemicals

£47.64 - £338.69

Chemical Identifiers

CAS 96-34-4
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00000931
InChI Key QABLOFMHHSOFRJ-UHFFFAOYSA-N
Synonym methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
PubChem CID 7295
IUPAC Name methyl 2-chloroacetate
SMILES COC(=O)CCl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10326600
View Documents
Thermo Scientific Acros
148630010
1 L Glass bottle
£47.64
1 L
Please to purchase this item. Need a web account? Register with us today!
10376690
View Documents
Thermo Scientific Acros
148630025
2.5 L Glass bottle
£101.51
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10036043
View Documents
Thermo Scientific
148630100
10 L Plastic drum
£338.69
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96-34-4
108.52
QABLOFMHHSOFRJ-UHFFFAOYSA-N
7295
COC(=O)CCl
C3H5ClO2
MFCD00000931
methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
methyl 2-chloroacetate

Specifications

96-34-4
Colorless
130.0°C to 132.0°C
Authentic
C3H5ClO2
1.4210 to 1.4230
02,197
1.23
Solubility in water: 28g/L (20°C). Other solubilities: soluble in methanol,ethanol,chloroform,,toluene,acetone,heptane,hexane,cyclohexane,,xylene and ether
COC(=O)CCl
108.52
1.23 mPa.s (20°C)
98%
Methyl chloroacetate
-33.0°C
1.2300g/mL
47°C
97.5% min. (GC)
ClCH2CO2CH3
MFCD00000931
15,6113
methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
QABLOFMHHSOFRJ-UHFFFAOYSA-N
methyl 2-chloroacetate
7295
108.52
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Toxic if swallowed.
May cause respiratory irritation.
Causes serious eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
Fatal in contact with skin.
May cause

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-501-1

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only