missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue, 1% w/v aq. soln.

Product Code. p-4408661
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11305597

Brand: Thermo Scientific Chemicals 042771.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122965-43-9 | C16H18ClN3S

Methylene Blue, 1% w/v aqueous solution is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It is also used in the sulfide analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylene Blue
CAS 122965-43-9, 7732-18-5
Assay Percent Range 1% w/v aqueous solution
Molecular Formula C16H18ClN3S
MDL Number MFCD00012111
Solubility Information Miscible with ethanol,chloroform,glacial acetic acid and glycerol. Slightly miscible with pyridine. Immiscible with ethyl ether,xylene,oleic acid,ethanol and acetone.
Formula Weight 319.86
Concentration or Composition (by Analyte or Components) 3, 7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate: 1.0%; Water: 99%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Quantity 100 mL
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.