missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Methylene Blue, high purity, biological stain Product Code.: 11443697

Thermo Scientific Chemicals Methylene Blue, high purity, biological stain

Product Code. 11443697
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11443697

Brand: Thermo Scientific Chemicals A18174.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methylene blue is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylene blue is used as a dye in different staining procedures viz. Wright′s stain and Jenner′s stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.

Solubility
Soluble in water, alcohol, acetic acid, glycerol and chloroform. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150006
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9; C.I. 52015
PubChem CID 16211647
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C

Specifications

Melting Point ∼180°C (decomposition)
Beilstein 3599847
Merck Index 14,6060
Solubility Information Soluble in water,alcohol,acetic acid,glycerol and chloroform. Insoluble in ether.
Formula Weight 319.86 (Anhydrous)
Quantity 25 g
Chemical Name or Material Methylene Blue, biological stain
Grade High purity

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252160 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog NumberA18174.14,A18174.36,A18174.22
Lot Number10252159 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog NumberA18174.14,A18174.36,A18174.22
Lot Number10250159 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberA18174.14,A18174.36,A18174.22
Lot Number10245673 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberA18174.14,A18174.36,A18174.22
Lot Number10241648 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog NumberA18174.14,A18174.36,A18174.22
compliance-icons
Product Identifier
  • Methylene Blue, high purity, biological stain
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Methylene Blue hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Methylene Blue, high purity, biological stain >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.