missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylene Blue hydrate, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
100g
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | 100.0°C to 110.0°C |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol |
| Infrared Spectrum | Authentic |
| IUPAC Name | 7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium chloride |
| Formula Weight | 319.85 |
| Loss on Drying | 22% max. |
| Percent Purity | ≥70% (TiCl3-titration) |
| Grade | Pure |
| Packaging | Plastic Drum |
| Color | Green |
| Show More |
Safety and Handling
Product Identifier
- Methylene Blue hydrate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- MIXTURE LIST-Contains : Methylene Blue hydrate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction