missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue, pure, certified, residual water

Product Code. p-163442 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10455081

Brand: Thermo Scientific Chemicals 414240250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Methylene Blue, C16H24ClN3O3S, CAS Number-7220-79-3, Basic Blue 9, 25g, 1.21 to 1.7, Vapor Pressure: Negligible, 27, V,19, 259, CHEBI:6872, Green, 13 (Air - 1.0), 82% min., 200-515-2, 319.85, Pure, Warning, GHS H Statement: Harmful if swallowed. | CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Basic Blue 9
PubChem CID 6099
ChEBI CHEBI:6872
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Melting Point 190.0°C
Absorption Ratio 1.21 to 1.7
Dye Content 82% min.
Beilstein 27, V,19, 259
Merck Index 15, 6132
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: in methanol: 430g/L
Infrared Spectrum Authentic
Lambda Max 663 to 667nm
Formula Weight 319.85
Grade Pure
Packaging Glass Bottle
Vapor Pressure Vapor Pressure: Negligible
Density 13 (Air = 1.0)
Color Green
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Methylene Blue
Show More Show Less
compliance-icons
Product Identifier
  • Methylene Blue
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.