Learn More
Methylene Blue trihydrate, Thermo Scientific Chemicals
Description
Methylene Blue trihydrate finds many uses in biology and chemistry. It acts as a GCS inhibitor, biological stain and redox indicator. It inhibits tau filament formation. Further, it is reported to bind to the M2 subtype of muscarinic receptors in rat cardiac myocytes and thus exhibit antimuscarinic effects on IK, ACh and ICa. It also inhibits endothelium-dependent relaxation of isolated blood vessels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methylene Blue trihydrate finds many uses in biology and chemistry. It acts as a GCS inhibitor, biological stain and redox indicator. It inhibits tau filament formation. Further, it is reported to bind to the M2 subtype of muscarinic receptors in rat cardiac myocytes and thus exhibit antimuscarinic effects on IK, ACh and ICa. It also inhibits endothelium-dependent relaxation of isolated blood vessels.
Solubility
Soluble in water, alcohol and chloroform. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
Beilstein | 3599847 |
Merck Index | 14,6060 |
Solubility Information | Soluble in water,alcohol and chloroform; Insoluble in ether. |
Formula Weight | 373.90 (319.86 Anhydrous) |
Odor | Odorless |
Color | Green |
Quantity | 25 g |
Physical Form | Crystalline Powder |
Chemical Name or Material | Methylene Blue trihydrate |
RUO – Research Use Only