missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Chloride, AR™, Macron Fine Chemicals™
178.60 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
2.5 L
Specifications
CAS | 75-09-2 |
Boiling Point | 39.8°C (1013 hPa) |
Quantity | 2.5 L |
Molecular Formula | CH2Cl2 |
MDL Number | MFCD00000881 |
UN Number | 1593 |
Synonym | Dichloromethane |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
SMILES | ClCCl |
IUPAC Name | dichloromethane |
Show More |
Safety and Handling
Signal Word
- Warning
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P281-Use personal protective equipment as required.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'