Learn More
Methylene di-p-phenyl diisocyanate, 98%, flakes, Thermo Scientific Chemicals
£29.75 - £43.22
Chemical Identifiers
CAS | 101-68-8, 5873-54-1 |
---|---|
Molecular Formula | C15H10N2O2 |
Molecular Weight (g/mol) | 250.26 |
MDL Number | MFCD00036131 |
InChI Key | UPMLOUAZCHDJJD-UHFFFAOYSA-N |
Synonym | 4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate |
PubChem CID | 7570 |
ChEBI | CHEBI:53218 |
IUPAC Name | 1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene |
SMILES | C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10055700
|
Thermo Scientific Acros
414281000 |
100 g | Glass bottle |
£29.75
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10371541
|
Thermo Scientific Acros
414285000 |
500 g | Glass bottle |
£43.22
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
101-68-8, 5873-54-1 | |
250.26 | |
UPMLOUAZCHDJJD-UHFFFAOYSA-N | |
7570 | |
1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene |
C15H10N2O2 | |
MFCD00036131 | |
4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate | |
CHEBI:53218 | |
C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O |
Specifications
101-68-8, 5873-54-1 | |
White | |
392.0°C | |
Authentic | |
C15H10N2O2 | |
MFCD00036131 | |
4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate | |
UPMLOUAZCHDJJD-UHFFFAOYSA-N | |
1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene | |
7570 | |
3-6 mPa.s (42°C) | |
98% | |
Methylene di-p-phenyl diisocyanate, 99.5% |
40.0°C | |
1.2200g/mL | |
202°C | |
2.5% max. (2,4′-MDI isomer) (GC) | |
OCNC6H4CH2C6H4NCO | |
1.22 | |
Solubility in water: decomposes. Other solubilities: soluble in acetone,benzene,nitrobenzene | |
C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O | |
250.26 | |
CHEBI:53218 | |
250.26 | |
Flakes |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Harmf
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-966-0
missing translation for 'rtecsNumber' : NQ9350000
missing translation for 'tsca' : TSCA
- Methylenedi-p-phenyl diisocyanate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
RUO – Research Use Only