missing translation for 'onlineSavingsMsg'
Learn More

Methyllithium, 1.6M sol. in diethyl ether (± 5% w/v), AcroSeal™, Thermo Scientific Chemicals

£336.68

Chemical Identifiers

CAS 917-54-4, 60-29-7
Molecular Formula CH3Li
Molecular Weight (g/mol) 21.98
MDL Number MFCD00008253
InChI Key DVSDBMFJEQPWNO-UHFFFAOYSA-N
Synonym methyllithium, lithium, methyl, methyl lithium, lithium methanide, meli, lithium methyl, lithium carbanide, lithium methide, methllithium, methyllithum
PubChem CID 2724049
SMILES [Li]C
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10386212
View Documents Promotion Details
Thermo Scientific Acros
188758000
800 mL AcroSeal™ Glass Bottle
£336.68
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

917-54-4, 60-29-7
21.98
DVSDBMFJEQPWNO-UHFFFAOYSA-N
2724049
CH3Li
MFCD00008253
methyllithium, lithium, methyl, methyl lithium, lithium methanide, meli, lithium methyl, lithium carbanide, lithium methide, methllithium, methyllithum
[Li]C

Specifications

Colorless to Yellow
CH3Li
MFCD00008253
0.7000g/mL
Solubility in water: reacts.
[Li]C
21.98
21.96
0.7
Solution
917-54-4, 60-29-7
CH3Li
01,686; 02,274; 03,202; 05,448; 06,384; 07,242; 08,342; 09,311; 11,340; 12,324; 16,219
methyllithium, lithium, methyl, methyl lithium, lithium methanide, meli, lithium methyl, lithium carbanide, lithium methide, methllithium, methyllithum
DVSDBMFJEQPWNO-UHFFFAOYSA-N
lithium;carbanide
2724049
4.5 to 5.5% (as MeLi)
−17°C
Methyllithium, Low Chloride, 1.6M Solution in Diethyl Ether (+/- 5% w/v)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Extremely flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Harmful if swallowed. Reacts violently with water. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Store in a dry place. Store in a closed container. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

missing translation for 'einecsNumber' : 213-026-4

missing translation for 'tsca' : TSCA

Product Identifier
  • Methyllithium, 1.6 M solution in diethyl ether

Signal Word
  • Danger

Hazard Category
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Lithium, methyl-, Ethyl ether

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only