missing translation for 'onlineSavingsMsg'
Learn More

Methyllycaconitine citrate, Tocris Bioscience™

Product Code. 30001166
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective α7 neuronal nAChR antagonist

Potent antagonist for α7-containing neuronal nicotinic receptors (Ki = 1.4 nM). Interacts with α4β2 and α6β2 receptors at concentrations > 40 nM. Attenuates METH-induced neurotoxicity in mouse striatum in vivo.

Chemical Identifiers

CAS 351344-10-0
Molecular Formula C43H58N2O17
Molecular Weight (g/mol) 874.934
InChI Key INBLZNJHDLEWPS-OXVGBIBJSA-N
Synonym methyllycaconitine citrate
PubChem CID 124081005
SMILES CCN1CC2(CCC(C34C2C(C(C31)(C5(CC(C6CC4C5C6OC)OC)O)O)OC)OC)COC(=O)C7=CC=CC=C7N8C(=O)CC(C8=O)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Quantity 25 mg
Formula Weight 874.93
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyllycaconitine citrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'