missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15401707
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylmagnesium bromide, 3M in ether has the wide range of applications in organic synthesis. Alkylation of aldehydes and ketones is the most common applications of Grignard reagent. It is a strong nucleophile that can form new carbon-carbon bonds. Also like organolithium compounds, it is useful to form carbon-heteroatom bonds.

Solubility
Soluble in ether, toluene and tetrahydrfuran. Insoluble in hydrocarbons.

Notes
It is highly flammable and reacts with moisture or water to form methane gas, so it should be stored in tightly closed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-16-1
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
PubChem CID 6349
SMILES C[Mg]Br

Specifications

Color Brown
Physical Form Liquid
Quantity 500 mL
Linear Formula CH3MgBr
UN Number UN1928
Flash Point -40°C (-40°F)
Synonym methyl magnesium bromide, grignard reagent
IUPAC Name bromo(methyl)magnesium
Formula Weight 119.26
Concentration or Composition (by Analyte or Components) 3M in ether
Odor Ether-like
Packaging ChemSeal™ bottles
Sensitivity Air and moisture sensitive
Density 1.035 g/mL
Chemical Name or Material Methylmagnesium bromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP16I008 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number041545.14,041545.36,041252.AE,041252.AP,041545.22
Lot NumberN21I013 Certificate TypeCertificate of Analysis Date30/06/2022 Catalog Number041545.14,041545.36,041252.AE,041252.AP,041545.22
Lot NumberY22H009 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog Number041545.14,041545.36,041252.AE,041252.AP,041545.22
Lot NumberL05S017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041545.14,041545.36,041252.AE,041252.AP,041545.22
Lot NumberI23U027 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041545.14,041545.36,041252.AE,041252.AP,041545.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.