missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
Click to view available options
500mL
Description
Methylmagnesium bromide is widely used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 75-16-1 |
Color | Brown to Colorless |
Physical Form | Liquid |
Quantity | 500 mL |
Molecular Formula | CH3BrMg |
Linear Formula | CH3MgBr |
MDL Number | MFCD00000041 |
UN Number | UN1928 |
Flash Point | -40°C (-40°F) |
Synonym | methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'