Learn More
Methylmagnesium iodide, 3M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
£359.79
Chemical Identifiers
CAS | 917-64-6, 60-29-7 |
---|---|
Molecular Formula | CH3IMg |
Molecular Weight (g/mol) | 166.24 |
MDL Number | MFCD00001026 |
InChI Key | AUPXBVDHVRZMIB-UHFFFAOYSA-M |
Synonym | methylmagnesium iodide, magnesium, iodomethyl, methylmagnesiumiodide, methyl magnesium iodide, iodomethylmagnesium, methylmagnesium iodide solution, 3.0 m in diethyl ether, memgi, ch3mgi, methyl-magnesium iodide |
PubChem CID | 101907 |
IUPAC Name | magnesium;carbanide;iodide |
SMILES | C[Mg]I |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10632445
|
Thermo Scientific Acros
427408000 |
800 mL | AcroSeal™ Glass Bottle |
£359.79
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
917-64-6, 60-29-7 | |
166.24 | |
AUPXBVDHVRZMIB-UHFFFAOYSA-M | |
101907 | |
C[Mg]I |
CH3IMg | |
MFCD00001026 | |
methylmagnesium iodide, magnesium, iodomethyl, methylmagnesiumiodide, methyl magnesium iodide, iodomethylmagnesium, methylmagnesium iodide solution, 3.0 m in diethyl ether, memgi, ch3mgi, methyl-magnesium iodide | |
magnesium;carbanide;iodide |
Specifications
917-64-6, 60-29-7 | |
−40°C | |
CH3MgI | |
1.26 | |
AUPXBVDHVRZMIB-UHFFFAOYSA-M | |
magnesium;carbanide;iodide | |
101907 | |
Methylmagnesium iodide |
1.2600g/mL | |
CH3IMg | |
MFCD00001026 | |
methylmagnesium iodide, magnesium, iodomethyl, methylmagnesiumiodide, methyl magnesium iodide, iodomethylmagnesium, methylmagnesium iodide solution, 3.0 m in diethyl ether, memgi, ch3mgi, methyl-magnesium iodide | |
C[Mg]I | |
166.24 | |
166.24 |
Safety and Handling
GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-031-1
- Methylmagnesium iodide, 3M solution in diethyl ether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H224-Extremely flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only