missing translation for 'onlineSavingsMsg'
Learn More

Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals

Product Code. 15471037
1 mL
Click to view available options
Quantity:
1 mL
10 mL
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

As a defoliant and epoxidation catalyst, as denaturing agent

Methyl mercury(II) hydroxide is used as defoliant and epoxidation catalyst, as denaturing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-57-2
MDL Number MFCD00013924

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 mL
Linear Formula CH3HgOH
UN Number UN2024
Formula Weight 232.63
Concentration or Composition (by Analyte or Components) 1M in H2O
Chemical Name or Material Methylmercury(II) hydroxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT08H040 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog Number013395.XF,013395.AD,013395.AB
Lot NumberU11J007 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number013395.XF,013395.AD,013395.AB

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.