missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylthymol Blue sodium salt
Click to view available options
Quantity:
5 g
Description
Methylthymol Blue sodium salt is used as thymol-containing organic dye. It is used as an indicator for complexometry. It is used in determination of sulfate in rainwater. It combines with silica gel and aids in the separation and preconcentration of trace amounts of heavy metals from some salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methylthymol Blue sodium salt is used as thymol-containing organic dye. It is used as an indicator for complexometry. It is used in determination of sulfate in rainwater. It combines with silica gel and aids in the separation and pr
Methylthymol Blue sodium salt is used as thymol-containing organic dye. It is used as an indicator for complexometry. It is used in determination of sulfate in rainwater. It combines with silica gel and aids in the separation and pr
Specifications
CAS | 1945-77-3 |
Molecular Formula | C37H44N2Na4O13S |
MDL Number | MFCD00148905 |
Synonym | Methylthymol Blue, water soluble |
Solubility Information | Soluble in water (460g/L) at 20°C. |
InChI Key | LGVVZVZPOQJZKT-UHFFFAOYSA-N |
SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] |
IUPAC Name | 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium |
Molecular Weight (g/mol) | 848.779 |
PubChem CID | 131850582 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'