missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylthymol blue, sodium salt , indicator grade
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Solubility Information | Solubility in water: 460g/L (20°C). |
Infrared Spectrum | Authentic |
Formula Weight | 844.76 |
Grade | ACS Reagent |
Packaging | Glass bottle |
Suitability | pH 6.5 (yellow) to pH 8.5 (blue):Pass Test |
Color | Red |
Quantity | 5 g |
Physical Form | Crystalline Powder |
Chemical Name or Material | Methylthymol blue, tetrasodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'