Learn More
Methyltin trichloride, 97%, Thermo Scientific Chemicals
£57.20 - £984.00
Chemical Identifiers
CAS | 993-16-8 |
---|---|
Molecular Formula | CH3Cl3Sn |
Molecular Weight (g/mol) | 240.095 |
MDL Number | MFCD00000514 |
InChI Key | YFRLQYJXUZRYDN-UHFFFAOYSA-K |
Synonym | methyltin trichloride, methyltrichlorotin, stannane, trichloromethyl, trichloromethyltin, monomethyltin trichloride, trichloro methyl stannane, methyltrichlorostannane, tin, methyl-, trichloride, unii-q2e6v14u06, ccris 6327 |
PubChem CID | 101940 |
IUPAC Name | trichloro(methyl)stannane |
SMILES | C[Sn](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334849
|
Thermo Scientific Alfa Aesar
071156.03 |
1 g | |||||||
11324849
|
Thermo Scientific Alfa Aesar
071156.06 |
5 g |
£224.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344849
|
Thermo Scientific Alfa Aesar
071156.14 |
25 g |
£984.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A chemical used in the synthesis of metallasilesesquiloxanes. Also used in the preparation of organotin(IV) complexes showing antibacterial activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
993-16-8 | |
240.095 | |
YFRLQYJXUZRYDN-UHFFFAOYSA-K | |
101940 | |
C[Sn](Cl)(Cl)Cl |
CH3Cl3Sn | |
MFCD00000514 | |
methyltin trichloride, methyltrichlorotin, stannane, trichloromethyl, trichloromethyltin, monomethyltin trichloride, trichloro methyl stannane, methyltrichlorostannane, tin, methyl-, trichloride, unii-q2e6v14u06, ccris 6327 | |
trichloro(methyl)stannane |
Specifications
993-16-8 | |
CH3Cl3Sn | |
UN2923 | |
methyltin trichloride, methyltrichlorotin, stannane, trichloromethyl, trichloromethyltin, monomethyltin trichloride, trichloro methyl stannane, methyltrichlorostannane, tin, methyl-, trichloride, unii-q2e6v14u06, ccris 6327 | |
YFRLQYJXUZRYDN-UHFFFAOYSA-K | |
trichloro(methyl)stannane | |
101940 | |
97% | |
Methyltin trichloride |
43°C | |
MFCD00000514 | |
Moisture sensitive | |
Soluble in Chloroform,Methanol. | |
C[Sn](Cl)(Cl)Cl | |
240.095 | |
240.11 | |
Crystalline |
Safety and Handling
GHS H Statement
H228-H300-H310-H330-H314-H318
Flammable solid.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H228-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 213-608-8
missing translation for 'rtecsNumber' : WH8585500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only