missing translation for 'onlineSavingsMsg'
Learn More
Methyltrioxorhenium(VII), 98%, Thermo Scientific Chemicals Product Code.: 11345005

Methyltrioxorhenium(VII), 98%, Thermo Scientific Chemicals

Product Code. 11345005
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 11345005

Brand: Thermo Scientific Alfa Aesar L18478.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Methyltrioxorhenium(VII) is used as a catalyst for various transformations such as olefin metathesis. It is associated with hydrogen peroxide and used as an oxidizing agent. It is used to prepare acid or ester from the corresponding terminalalkynes. It is a catalytic oxidant used with urea hydrogen peroxide to prepare nitrones. It reacts with 2-methylnaphthalene to get 2-methyl-1,4-naphthoquinone (Vitamin K3). Further, it catalyzes the reaction with aldehydes and diazoketones to get an alkene. In addition to this, it is used as an effective antioxidant under various conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltrioxorhenium(VII) is used as a catalyst for various transformations such as olefin metathesis. It is associated with hydrogen peroxide and used as an oxidizing agent. It is used to prepare acid or ester from the corresponding terminalalkynes. It is a catalytic oxidant used with urea hydrogen peroxide to prepare nitrones. It reacts with 2-methylnaphthalene to get 2-methyl-1,4-naphthoquinone (Vitamin K3). Further, it catalyzes the reaction with aldehydes and diazoketones to get an alkene. In addition to this, it is used as an effective antioxidant under various conditions.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70197-13-6
Molecular Formula CH3O3Re-
Molecular Weight (g/mol) 249.239
MDL Number MFCD00192332
InChI Key VZSXFJPZOCRDPW-UHFFFAOYSA-N
Synonym methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide viiShow MoreShow Less
PubChem CID 2734010
IUPAC Name carbanide;trioxorhenium
SMILES [CH3-].O=[Re](=O)=O

Specifications

Melting Point 110°C to 115°C
Quantity 100 mg
Linear Formula CH3ReO3
UN Number UN1479
Beilstein 3927932
Solubility Information Soluble in water.
Formula Weight 249.24
Percent Purity 98%
Sensitivity Light and moisture sensitive
Chemical Name or Material Methyltrioxorhenium(VII)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC19S025 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL18478.ME,L18478.MC
Lot NumberL27Q014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL18478.ME,L18478.MC
Lot Number10189519 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL18478.ME,L18478.MC
Lot NumberL08Q009 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL18478.ME,L18478.MC
Lot NumberR07A013 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL18478.ME,L18478.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyltrioxorhenium(VII), 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.