missing translation for 'onlineSavingsMsg'
Learn More

Methylurea, 97%, Thermo Scientific Chemicals

Product Code. 10001360
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-50-5
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.08
MDL Number MFCD00007950
InChI Key XGEGHDBEHXKFPX-UHFFFAOYSA-N
Synonym 1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
PubChem CID 11719
ChEBI CHEBI:44383
IUPAC Name methylurea
SMILES CNC(=O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 98.0°C to 102.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (HPLC)
Packaging Plastic bottle
Linear Formula CH3NHCONH2
Quantity 100 g
Beilstein 04, 64
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: freely soluble in ethanol
Formula Weight 74.08
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material Methylurea, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'