missing translation for 'onlineSavingsMsg'
Learn More

Metolazone, Fisher BioReagents

Product Code. 10134624
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Packaging:
Amber Glass
Unit Size:
100mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 10134624

Brand: Fisher BioReagents BP2667100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Metolazone is an anti-hypertensive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17560-51-9
Molecular Formula C16H16ClN3O3S
Molecular Weight (g/mol) 365.83
MDL Number MFCD00069304
InChI Key AQCHWTWZEMGIFD-UHFFFAOYNA-N
Synonym metolazone, zaroxolyn, diulo, mykrox, microx, metalozone, metenix, oldren, metolazona, metolazonum
PubChem CID 4170
ChEBI CHEBI:64354
SMILES CC1NC2=CC(Cl)=C(C=C2C(=O)N1C1=CC=CC=C1C)S(N)(=O)=O

Specifications

Melting Point 248°C
Color White
Packaging Amber Glass
Quantity 100 mg
Merck Index 15, 6224
IUPAC Name 7-chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2-dihydroquinazoline-6-sulfonamide
Formula Weight 365.8
Physical Form Powder or Solid
Chemical Name or Material Metolazone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.